Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
423608
Gene Name
WAPL cohesin release factor
Gene Aliases
WAPAL
Chromosome Location
Chr.6: 2968226 - 3064459 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199511.1 NP_001186440.1 8 - 9 3117 96
XM_015287975.2 XP_015143461.2 11 - 12 3030 96
XM_015287976.2 XP_015143462.1 9 - 10 2744 96
XM_015287977.2 XP_015143463.1 9 - 10 2572 96
XM_015287978.2 XP_015143464.2 8 - 9 2392 96
XM_025151506.1 XP_025007274.1 9 - 10 2441 96
GenBank mRNA BX929506.1 - 1 - 2 162 96

View Full Product Details