Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
423608
Gene Name
WAPL cohesin release factor
Gene Aliases
WAPAL
Chromosome Location
Chr.6: 2968226 - 3064459 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199511.1 NP_001186440.1 14 - 15 3856 86
XM_015287975.2 XP_015143461.2 17 - 18 3769 86
XM_015287976.2 XP_015143462.1 15 - 16 3483 86
XM_015287977.2 XP_015143463.1 15 - 16 3311 86
XM_015287978.2 XP_015143464.2 14 - 15 3131 86
XM_025151506.1 XP_025007274.1 15 - 16 3180 86
GenBank mRNA BX929506.1 - 7 - 8 901 86

View Full Product Details