Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT15
Entrez Gene ID
418858
Gene Name
nudix hydrolase 15
Gene Aliases
-
Chromosome Location
Chr.1: 169995439 - 169998554 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_417053.6 XP_417053.4 2 - 3 2042 146
GenBank mRNA BX929709.1 - 2 - 3 396 146
CR386374.1 - 2 - 3 403 146

View Full Product Details