Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC51B
Entrez Gene ID
415531
Gene Name
solute carrier family 51 beta subunit
Gene Aliases
OSTBETA
Chromosome Location
Chr.10: 18212671 - 18221175 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015292256.2 XP_015147742.1 4 - 5 1082 71
XM_025153900.1 XP_025009668.1 4 - 5 765 71
GenBank mRNA BX929774.1 - 5 - 6 893 71
BX930136.2 - 3 - 4 873 71

View Full Product Details