Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPCS1
Entrez Gene ID
415899
Gene Name
signal peptidase complex subunit 1
Gene Aliases
-
Chromosome Location
Chr.12: 1180932 - 1183221 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171644.1 NP_001165115.1 1 - 2 272 78
NM_001171645.1 NP_001165116.1 1 - 2 272 78
GenBank mRNA BX929433.1 - 1 - 2 220 78
BX929751.2 - 1 - 2 74 78
BX929814.1 - 1 - 2 231 78
BX931945.1 - 1 - 2 74 78
BX932880.1 - 1 - 2 212 78
BX933705.1 - 1 - 2 220 78

View Full Product Details