Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD49
Entrez Gene ID
418997
Gene Name
ankyrin repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.1: 186932542 - 186935338 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003640591.4 XP_003640639.1 2 - 3 325 78
XM_417191.5 XP_417191.1 2 - 3 333 78
GenBank mRNA BX932571.1 - 1 - 2 292 78
BX932620.1 - 1 - 2 292 78
BX933170.1 - 2 - 3 307 78
BX935515.2 - 2 - 3 321 78

View Full Product Details