Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIEZO2
Entrez Gene ID
421052
Gene Name
piezo type mechanosensitive ion channel component 2
Gene Aliases
FAM38B
Chromosome Location
Chr.2: 97719562 - 98025242 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015282541.2 XP_015138027.2 11 - 12 1768 72
XM_015282543.2 XP_015138029.2 11 - 12 1768 72
XM_025147818.1 XP_025003586.1 11 - 12 1768 72
XM_025147819.1 XP_025003587.1 10 - 11 1656 72
XM_025147820.1 XP_025003588.1 11 - 12 1768 72
XM_025147821.1 XP_025003589.1 7 - 8 1131 72
XM_025147822.1 XP_025003590.1 11 - 12 1768 72
XM_025147824.1 XP_025003592.1 11 - 12 1768 72
GenBank mRNA BX933406.1 - 3 - 4 539 72

View Full Product Details