Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDHB
Entrez Gene ID
416066
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Gene Aliases
-
Chromosome Location
Chr.12: 12225248 - 12228987 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198620.1 NP_001185549.1 7 - 8 797 103
GenBank mRNA BX929583.1 - 6 - 7 698 103
BX933955.1 - 7 - 8 797 103

View Full Product Details