Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C10H15ORF48
Entrez Gene ID
415442
Gene Name
chromosome 10 open reading frame, human C15orf48
Gene Aliases
C15ORF48
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX931191.1 - 3 - 4 338 122
BX934528.1 - 3 - 4 316 122

View Full Product Details