Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM114A2
Entrez Gene ID
416262
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
C5orf3
Chromosome Location
Chr.13: 12979733 - 12989395 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_414580.5 XP_414580.1 6 - 7 739 79
GenBank mRNA BX934896.1 - 3 - 4 157 79

View Full Product Details