Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTD
Entrez Gene ID
420639
Gene Name
biotinidase
Gene Aliases
-
Chromosome Location
Chr.2: 34016958 - 34027627 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199624.1 NP_001186553.1 3 - 4 435 72
GenBank mRNA BX935121.1 - 3 - 4 406 72
CN235934.1 - 3 - 4 435 72
CN236789.1 - 3 - 4 434 72

View Full Product Details