Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
418180
Gene Name
RERG like
Gene Aliases
-
Chromosome Location
Chr.1: 63888788 - 63899627 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_416411.6 XP_416411.1 4 - 5 474 118
GenBank mRNA BX950371.1 - 4 - 5 476 118

View Full Product Details