Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATSL2
Entrez Gene ID
417483
Gene Name
GATS protein like 2
Gene Aliases
CASTOR2, GATS, GATSL1
Chromosome Location
Chr.19: 2690346 - 2784509 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281807.1 NP_001268736.1 8 - 9 1238 85
XM_015295702.2 XP_015151188.1 9 - 10 1471 85
XM_025141933.1 XP_024997701.1 9 - 10 1773 85
XM_025141934.1 XP_024997702.1 8 - 9 1386 85
GenBank mRNA BX950387.1 - 8 - 9 988 85
BX950514.1 - 8 - 9 922 85
CR405722.1 - 8 - 9 1032 85

View Full Product Details