Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF830
Entrez Gene ID
420966
Gene Name
zinc finger protein 830
Gene Aliases
CCDC16
Chromosome Location
Chr.2: 85614485 - 85626086 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419050.6 XP_419050.1 6 - 7 754 66
GenBank mRNA BX930669.1 - 2 - 3 284 66
BX936222.2 - 6 - 7 720 66
BX936269.2 - 6 - 7 720 66
BX950392.1 - 6 - 7 720 66

View Full Product Details