Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSPH9
Entrez Gene ID
421446
Gene Name
radial spoke head component 9
Gene Aliases
C6orf206
Chromosome Location
Chr.3: 30921534 - 30942828 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277604.1 NP_001264533.1 1 - 2 246 133
XM_015283712.2 XP_015139198.1 1 - 2 222 133
GenBank mRNA BX950488.1 - 1 - 2 246 133
CR407494.1 - 1 - 2 216 133

View Full Product Details