Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST6GALNAC5
Entrez Gene ID
407778
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
SIAT7E
Chromosome Location
Chr.8: 19606479 - 19722947 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277494.1 NP_001264423.1 5 - 6 868 119
XM_025152775.1 XP_025008543.1 4 - 5 1172 119
GenBank mRNA AJ646877.1 - 5 - 6 770 119

View Full Product Details