Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP12
Entrez Gene ID
421634
Gene Name
A-kinase anchoring protein 12
Gene Aliases
-
Chromosome Location
Chr.3: 48914530 - 48945833 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004935624.3 XP_004935681.1 1 - 2 256 69
GenBank mRNA BX933212.2 - 1 - 2 34 69

View Full Product Details