Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXNL2
Entrez Gene ID
426175
Gene Name
nucleoredoxin-like 2
Gene Aliases
-
Chromosome Location
Chr.Z: 43280129 - 43302837 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX933348.2 - 1 - 2 455 98

View Full Product Details