Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAT1A
Entrez Gene ID
423628
Gene Name
methionine adenosyltransferase 1A
Gene Aliases
-
Chromosome Location
Chr.6: 5149809 - 5166476 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199519.1 NP_001186448.1 4 - 5 626 73
GenBank mRNA BX933611.1 - 4 - 5 626 73
BX935026.2 - 4 - 5 526 73
BX935281.1 - 4 - 5 626 73
BX935287.2 - 4 - 5 626 73

View Full Product Details