Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2E
Entrez Gene ID
420103
Gene Name
RNA polymerase II subunit E
Gene Aliases
-
Chromosome Location
Chr.28: 3032175 - 3034544 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_418224.6 XP_418224.2 4 - 5 469 107
GenBank mRNA BX929663.1 - 4 - 5 453 107
BX930905.2 - 4 - 5 452 107
BX935423.2 - 4 - 5 456 107
BX935442.2 - 4 - 5 444 107
CR385484.1 - 4 - 5 432 107

View Full Product Details