Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERICH1
Entrez Gene ID
421904
Gene Name
glutamate rich 1
Gene Aliases
-
Chromosome Location
Chr.3: 91475835 - 91569878 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278029.1 NP_001264958.1 1 - 2 109 61
XM_015284836.2 XP_015140322.1 1 - 2 61
XM_015284837.2 XP_015140323.2 1 - 2 61
GenBank mRNA BU328393.1 - 1 - 2 109 61
BX930034.1 - 1 - 2 72 61
BX935781.2 - 1 - 2 34 61

View Full Product Details