Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DZANK1
Entrez Gene ID
416722
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
C20orf12, C3H20orf12
Chromosome Location
Chr.3: 15498922 - 15519217 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199396.1 NP_001186325.1 17 - 18 1955 93
XM_015283472.2 XP_015138958.1 17 - 18 2059 93
XM_015283473.2 XP_015138959.1 17 - 18 2059 93
XM_025148594.1 XP_025004362.1 15 - 16 1866 93
XM_025148595.1 XP_025004363.1 14 - 15 1587 93
GenBank mRNA CR353555.1 - 8 - 9 859 93

View Full Product Details