Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSL1D1
Entrez Gene ID
416416
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 510063 - 515427 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039263.2 NP_001034352.2 4 - 5 487 139
XM_015294144.2 XP_015149630.1 4 - 5 480 139
GenBank mRNA AJ456913.1 - 4 - 5 488 139
CR387393.1 - 4 - 5 474 139

View Full Product Details