Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LETM2
Entrez Gene ID
426779
Gene Name
leucine zipper and EF-hand containing transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.22: 2637207 - 2642017 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025142904.1 XP_024998672.1 2 - 3 187 69
XM_025142905.1 XP_024998673.1 4 - 5 950 69
GenBank mRNA CR387422.1 - 4 - 5 860 69
CR523710.1 - 4 - 5 629 69

View Full Product Details