Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D31
Entrez Gene ID
420349
Gene Name
TBC1 domain family member 31
Gene Aliases
WDR67
Chromosome Location
Chr.2: 137916084 - 137941378 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015283159.2 XP_015138645.1 18 - 19 2610 117
XM_025148148.1 XP_025003916.1 17 - 18 2982 117
XM_025148149.1 XP_025003917.1 17 - 18 2982 117
XM_025148150.1 XP_025003918.1 17 - 18 2494 117
XM_418456.6 XP_418456.2 17 - 18 2585 117
GenBank mRNA CR387563.1 - 3 - 4 377 117

View Full Product Details