Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL26A1
Entrez Gene ID
417504
Gene Name
collagen type XXVI alpha 1 chain
Gene Aliases
EMID2
Chromosome Location
Chr.19: 3659652 - 3832077 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004946472.3 XP_004946529.1 5 - 6 759 65
XM_015295986.2 XP_015151472.1 6 - 7 802 65
XM_015295987.2 XP_015151473.1 6 - 7 796 65
XM_015295988.2 XP_015151474.1 6 - 7 802 65
XM_015295989.2 XP_015151475.1 6 - 7 801 65
XM_015295990.2 XP_015151476.1 5 - 6 753 65
XM_015295991.2 XP_015151477.1 6 - 7 627 65
XM_015295993.2 XP_015151479.1 6 - 7 653 65
GenBank mRNA CR387598.1 - 5 - 6 536 65

View Full Product Details