Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL2
Entrez Gene ID
416874
Gene Name
myosin, light chain 2, regulatory, cardiac, slow
Gene Aliases
G2, MLC-2A, MLC2
Chromosome Location
Chr.15: 6330267 - 6334277 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271929.1 NP_001258858.1 1 - 2 73 60
GenBank mRNA BU257651.1 - 1 - 2 78 60
CR387719.1 - 1 - 2 217 60

View Full Product Details