Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS11
Entrez Gene ID
415498
Gene Name
mitochondrial ribosomal protein S11
Gene Aliases
-
Chromosome Location
Chr.10: 13612326 - 13615857 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005821.1 NP_001005821.1 3 - 4 282 76
GenBank mRNA AJ719914.1 - 3 - 4 282 76

View Full Product Details