Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP1
Entrez Gene ID
420540
Gene Name
STEAP family member 1
Gene Aliases
-
Chromosome Location
Chr.2: 21641329 - 21651293 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015281969.1 XP_015137455.1 4 - 5 863 136
GenBank mRNA BX929749.2 - 1 - 2 142 136

View Full Product Details