Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAT2
Entrez Gene ID
421670
Gene Name
adenosine deaminase, tRNA specific 2
Gene Aliases
DEADC1
Chromosome Location
Chr.3: 52558887 - 52568436 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419709.6 XP_419709.1 1 - 2 138 63
GenBank mRNA BX930226.2 - 1 - 2 127 63

View Full Product Details