Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GXYLT2
Entrez Gene ID
416097
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
GLT8D4
Chromosome Location
Chr.12: 16898367 - 16917475 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015293228.2 XP_015148714.1 5 - 6 147
XM_015293229.2 XP_015148715.1 5 - 6 147
GenBank mRNA BX931958.2 - 5 - 6 875 147

View Full Product Details