Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3GALNT1
Entrez Gene ID
425011
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
B3GALT3
Chromosome Location
Chr.9: 22252415 - 22276447 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025153494.1 XP_025009262.1 1 - 2 143
XM_025153495.1 XP_025009263.1 1 - 2 143
XM_025153497.1 XP_025009265.1 1 - 2 143
XM_025153499.1 XP_025009267.1 1 - 2 143
XM_025153501.1 XP_025009269.1 1 - 2 143
GenBank mRNA CR387037.1 - 1 - 2 295 143

View Full Product Details