Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERI3
Entrez Gene ID
424579
Gene Name
ERI1 exoribonuclease family member 3
Gene Aliases
PRNPIP
Chromosome Location
Chr.8: 20806975 - 20902966 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015291303.1 XP_015146789.1 2 - 3 307 77
XM_015291304.2 XP_015146790.1 2 - 3 292 77
XM_015291305.2 XP_015146791.1 2 - 3 411 77
GenBank mRNA CR389892.1 - 1 - 2 282 77

View Full Product Details