Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM117B
Entrez Gene ID
424088
Gene Name
family with sequence similarity 117 member B
Gene Aliases
ALS2CR13
Chromosome Location
Chr.7: 11930105 - 11958813 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942631.3 XP_004942688.2 5 - 6 1224 109
XM_421944.6 XP_421944.5 5 - 6 1474 109
GenBank mRNA CR390643.1 - 1 - 2 137 109

View Full Product Details