Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2D6
Entrez Gene ID
417981
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CYP2D49, CYP2D6c
Chromosome Location
Chr.1: 49408430 - 49417609 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195557.1 NP_001182486.1 9 - 9 2728 116
GenBank mRNA CR354312.1 - 9 - 9 2728 116
CR406594.1 - 1 - 1 1050 116

View Full Product Details