Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STKLD1
Entrez Gene ID
417154
Gene Name
serine/threonine kinase like domain containing 1
Gene Aliases
C17H9ORF96, C9orf96
Chromosome Location
Chr.17: 7189460 - 7201770 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004945935.3 XP_004945992.1 14 - 15 1986 66
XM_004945936.3 XP_004945993.1 14 - 15 1983 66
XM_004945937.3 XP_004945994.1 13 - 14 1882 66
XM_004945938.3 XP_004945995.1 13 - 14 1855 66
XM_004945939.3 XP_004945996.1 14 - 15 1706 66
XM_025141407.1 XP_024997175.1 13 - 14 1851 66
XM_025141408.1 XP_024997176.1 14 - 15 1700 66
GenBank mRNA CR523822.1 - 3 - 4 356 66

View Full Product Details