Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL35A
Entrez Gene ID
424924
Gene Name
ribosomal protein L35a
Gene Aliases
-
Chromosome Location
Chr.9: 15278279 - 15281119 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277457.1 NP_001264386.1 3 - 4 249 97
XM_025153370.1 XP_025009138.1 3 - 4 259 97
GenBank mRNA BU254781.1 - 3 - 4 218 97
BU255532.1 - 3 - 4 215 97
BU328575.1 - 3 - 4 249 97
CR390693.1 - 2 - 3 476 97
CR405821.1 - 3 - 4 216 97
CR524410.1 - 3 - 4 227 97

View Full Product Details