The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167703.1 | NP_001161175.1 | 6 - 7 | 895 | 99 |
NM_001167704.1 | NP_001161176.1 | 6 - 7 | 895 | 99 | |
GenBank mRNA | X56647.1 | - | - | 895 | 99 |
X56648.1 | - | - | 598 | 99 |