Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRRD
Entrez Gene ID
416908
Gene Name
SRR1 domain containing
Gene Aliases
-
Chromosome Location
Chr.15: 7445241 - 7447016 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ720235.1 - 1 - 2 250 126

View Full Product Details