Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP7B1
Entrez Gene ID
420164
Gene Name
cytochrome P450 family 7 subfamily B member 1
Gene Aliases
-
Chromosome Location
Chr.2: 114389502 - 114513649 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025147742.1 XP_025003510.1 4 - 5 1501 68
GenBank mRNA AY700579.1 - 4 - 5 1025 68

View Full Product Details