Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PURA
- Entrez Gene ID
-
430546
- Gene Name
- purine rich element binding protein A
- Gene Aliases
-
PURB
- Chromosome Location
-
Chr.13: 2072754 - 2124524 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR354163.1 |
- |
3 - 4 |
268 |
138 |
- Gene Symbol
- CYSTM1
- Entrez Gene ID
-
769167
- Gene Name
- cysteine rich transmembrane module containing 1
- Gene Aliases
-
C13H5orf32, C5orf32
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR389441.1 |
- |
2 - 3 |
356 |
138 |
View Full Product Details