Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC51
Entrez Gene ID
416045
Gene Name
coiled-coil domain containing 51
Gene Aliases
-
Chromosome Location
Chr.12: 11664463 - 11669126 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004944615.3 XP_004944672.1 4 - 5 832 58
XM_414385.6 XP_414385.2 4 - 5 1019 58
GenBank mRNA CR405782.1 - 2 - 3 445 58

View Full Product Details