Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP4R1L
Entrez Gene ID
419319
Gene Name
protein phosphatase 4, regulatory subunit 1-like
Gene Aliases
-
Chromosome Location
Chr.20: 11230671 - 11251485 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ851434.1 - 18 - 19 2783 123
CR406377.1 - 5 - 6 630 123

View Full Product Details