Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPHOSPH6
Entrez Gene ID
415811
Gene Name
M-phase phosphoprotein 6
Gene Aliases
-
Chromosome Location
Chr.11: 15802151 - 15809325 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277139.1 NP_001264068.1 3 - 4 348 152
GenBank mRNA BX929849.2 - 3 - 4 241 152
BX930843.2 - 3 - 4 257 152
BX935551.1 - 3 - 4 347 152
BX936086.2 - 3 - 4 316 152
CR406488.1 - 3 - 4 241 152

View Full Product Details