Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENKUR
Entrez Gene ID
420498
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
C10orf63
Chromosome Location
Chr.2: 16731138 - 16740294 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR407347.1 - 2 - 3 227 110

View Full Product Details