Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTBP3
Entrez Gene ID
427333
Gene Name
polypyrimidine tract binding protein 3
Gene Aliases
ROD1
Chromosome Location
Chr.Z: 66269909 - 66324686 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004949515.3 XP_004949572.1 7 - 8 778 112
XM_004949517.3 XP_004949574.1 6 - 7 657 112
XM_015280701.1 XP_015136187.1 7 - 8 830 112
XM_015280702.1 XP_015136188.1 8 - 9 979 112
XM_015280703.2 XP_015136189.1 7 - 8 776 112
XM_015280704.2 XP_015136190.1 7 - 8 979 112
XM_025144918.1 XP_025000686.1 6 - 7 709 112
XM_424912.6 XP_424912.3 7 - 8 740 112
GenBank mRNA CR524084.1 - 5 - 6 572 112

View Full Product Details