Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
416229
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
Chromosome Location
Chr.13: 11124638 - 11127201 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277714.1 NP_001264643.1 3 - 4 553 119
GenBank mRNA BX929782.1 - 3 - 4 553 119

View Full Product Details