The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_422088.5 | XP_422088.1 | 2 - 3 | 283 | 105 |
GenBank mRNA | BX930757.1 | - | 2 - 3 | 262 | 105 |