Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPBP1L1
Entrez Gene ID
424603
Gene Name
GC-rich promoter binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.8: 21542362 - 21574445 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277583.1 NP_001264512.1 5 - 6 668 68
XM_015291008.2 XP_015146494.1 6 - 7 1581 68
GenBank mRNA BX931765.1 - 4 - 5 587 68
CR353506.1 - 5 - 6 668 68

View Full Product Details