Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBP2
Entrez Gene ID
424822
Gene Name
retinol binding protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 6067154 - 6078035 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277417.1 NP_001264346.1 3 - 4 108
GenBank mRNA BU296446.1 - 3 - 4 388 108
BX930467.1 - 3 - 4 410 108
BX930583.2 - 3 - 4 408 108
BX932676.1 - 3 - 4 427 108
CD735799.1 - 1 - 2 108

View Full Product Details